KEY407051062 Key Organics / BIONET

[2-(benzyldimethylsilyl)phenyl]methanol

  • MDL number MFCD20922837
  • Cas Number : 201050-00-2
  • Molecular Weight : 256.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362705

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2248_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart