KEY407051103 Key Organics / BIONET

N-[(oxolan-2-yl)methyl]-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD09266189
  • Cas Number : 1073355-06-2
  • Molecular Weight : 331.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362746

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2292_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart