KEY407051158 Key Organics / BIONET

(E)-benzyl({[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylidene})amine

  • MDL number MFCD23381612
  • Cas Number : 2097555-05-8
  • Molecular Weight : 321.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362801

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2350_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart