KEY407051184 Key Organics / BIONET

potassium trifluoro[4-(piperidine-1-carbonyl)phenyl]boranuide

  • MDL number MFCD11977715
  • Cas Number : 1359865-98-7
  • Molecular Weight : 295.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362827

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2383_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart