KEY407051195 Key Organics / BIONET

4,4,5,5-tetramethyl-2-[6-(2-methylpropoxy)naphthalen-2-yl]-1,3,2-dioxaborolane

  • MDL number MFCD26142761
  • Cas Number : 1363386-57-5
  • Molecular Weight : 326.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362838

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2396_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart