KEY407051198 Key Organics / BIONET

8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline

  • MDL number MFCD05663894
  • Cas Number : 190788-62-6
  • Molecular Weight : 255.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Inchi Code 1S/C15H18BNO2/c1-14(2)15(3,4)19-16(18-14)12-9-5-7-11-8-6-10-17-13(11)12/h5-10H,1-4H3
Inchi Key RNTGVJBKPQOGPS-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store under N2, Hygroscopic
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362841

Safety Information

signalword Warning
pcodes P232,P260,P270,P280,P402 + P404,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2399_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart