KEY407051203 Key Organics / BIONET

1,3-dimethyl-5-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylidene}-1,3-diazinane-2,4,6-trione

  • MDL number MFCD20527089
  • Cas Number : 2027543-16-2
  • Molecular Weight : 370.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362846

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2404_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart