KEY407051216 Key Organics / BIONET

2-(3-ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD16294529
  • Cas Number : 946168-04-3
  • Molecular Weight : 228.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362859

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2418_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart