KEY407051241 Key Organics / BIONET

2-(9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD16294548
  • Cas Number : 922706-40-9
  • Molecular Weight : 292.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Inchi Code 1S/C19H21BO2/c1-18(2)19(3,4)22-20(21-18)15-9-10-17-14(12-15)11-13-7-5-6-8-16(13)17/h5-10,12H,11H2,1-4H3
Inchi Key WVJQQLZHOADEAF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362884

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2449_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart