KEY407051258 Key Organics / BIONET

N-cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD05663848
  • Cas Number : 827614-68-6
  • Molecular Weight : 287.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Inchi Code 1S/C16H22BNO3/c1-15(2)16(3,4)21-17(20-15)12-7-5-11(6-8-12)14(19)18-13-9-10-13/h5-8,13H,9-10H2,1-4H3,(H,18,19)
Inchi Key PXTNJXLWXIJJIM-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362901

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2475_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart