KEY407051269 Key Organics / BIONET

potassium (4-butylphenyl)trifluoroboranuide

  • MDL number MFCD18861573
  • Cas Number : 1412414-09-5
  • Molecular Weight : 240.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362912

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2494_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart