KEY407051286 Key Organics / BIONET

4,4,5,5-tetramethyl-2-(2-methylphenyl)-1,3,2-dioxaborolane

  • MDL number MFCD06795658
  • Cas Number : 195062-59-0
  • Molecular Weight : 218.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362929

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2518_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart