KEY407051288 Key Organics / BIONET

[7-(dihydroxyboranyl)-9,9-dimethyl-9H-fluoren-2-yl]boronic acid

  • MDL number MFCD08704228
  • Cas Number : 866100-14-3
  • Molecular Weight : 281.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362931

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2520_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart