KEY407051318 Key Organics / BIONET

4-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}morpholine hydrochloride

  • MDL number MFCD06657894
  • Cas Number : 944591-57-5
  • Molecular Weight : 339.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362961

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2564_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart