KEY407051328 Key Organics / BIONET

potassium trifluoro(naphthalen-1-yl)boranuide

  • MDL number MFCD06659936
  • Cas Number : 166328-07-0
  • Molecular Weight : 234.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362971

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2577_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart