KEY407051352 Key Organics / BIONET

2-[4-(2-methoxyethoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD23143157
  • Cas Number : 959972-40-8
  • Molecular Weight : 278.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362995

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2603_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart