KEY407051364 Key Organics / BIONET

2-(3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD18375236
  • Cas Number : 936618-92-7
  • Molecular Weight : 222.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Inchi Code 1S/C12H16BFO2/c1-11(2)12(3,4)16-13(15-11)9-6-5-7-10(14)8-9/h5-8H,1-4H3
Inchi Key VWBPPYBBVMPZOZ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/363007

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2619_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart