KEY407051378 Key Organics / BIONET

4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-carbaldehyde

  • MDL number MFCD18447531
  • Cas Number : 1040424-52-9
  • Molecular Weight : 308.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/363021

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2633_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart