KEY407051400 Key Organics / BIONET

N-ethyl-N-(propan-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • MDL number MFCD26142768
  • Molecular Weight : 289.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/363043

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2659_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart