KEY407051408 Key Organics / BIONET

methyl 3-acetamido-4-(methylamino)benzoate

  • MDL number MFCD26142759
  • Cas Number : 256936-10-4
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151 - 153
Purity 97
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link http://www.keyorganics.net/bionet/catalog/product/view/id/363055

Safety Information

signalword Warning
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart