KEY408584496 Key Organics / BIONET

N-cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • MDL number MFCD26383468
  • Cas Number : 1430474-31-9
  • Molecular Weight : 287.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/363085

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2664_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart