KEY408584511 Key Organics / BIONET

N'-tert-butyl(tert-butoxy)carbohydrazide

  • MDL number MFCD04125557
  • Cas Number : 60295-52-5
  • Molecular Weight : 188.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76 - 79
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77364

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0858_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart