KEY408584512 Key Organics / BIONET

2-(anthracen-9-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD24039741
  • Cas Number : 709022-63-9
  • Molecular Weight : 304.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/363086

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2666_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart