KEY408584523 Key Organics / BIONET

potassium (3,5-dibromophenyl)trifluoroboranuide

  • MDL number MFCD26383469
  • Cas Number : 929626-21-1
  • Molecular Weight : 341.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/363097

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2688_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart