KEY408584526 Key Organics / BIONET

potassium trifluoro(4-methanesulfonylphenyl)boranuide

  • MDL number MFCD04115738
  • Cas Number : 850623-40-4
  • Molecular Weight : 262.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/363100

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2692_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart