KEY408584535 Key Organics / BIONET

(1S,2R,5R)-3-[(tert-butoxy)carbonyl]-3-azabicyclo[3.1.0]hexane-2-carboxylic acid

  • MDL number MFCD11042445
  • Cas Number : 1051393-66-8
  • Molecular Weight : 227.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133 - 134
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97534

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GF-0238_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart