KEY410536871 Key Organics / BIONET

N'-(2-methyl-1-nitropropan-2-yl)(tert-butoxy)carbohydrazide

  • MDL number MFCD26523301
  • Cas Number : 1354825-84-5
  • Molecular Weight : 233.268

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/367316

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2718_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart