KEY410536877 Key Organics / BIONET

4-methyl-6-nitro-2H-1,3-benzodioxole-5-carboxylic acid

  • MDL number MFCD26523308
  • Cas Number : 861588-35-4
  • Molecular Weight : 225.156

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368085

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4648_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart