KEY410536879 Key Organics / BIONET

5-bromo-3-(2-methoxyphenyl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD26523310
  • Cas Number : 875639-78-4
  • Molecular Weight : 303.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368094

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4657_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart