KEY410536881 Key Organics / BIONET

tert-butyl(2-oxo-3-phenoxypropyl)amine hydrochloride

  • MDL number MFCD26523313
  • Cas Number : 343319-03-9
  • Molecular Weight : 257.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368099

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4664_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart