KEY410536893 Key Organics / BIONET

(3,4-diaminophenyl)methanol

  • MDL number MFCD00173951
  • Cas Number : 63189-98-0
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97 - 99
Purity 90
Storage temp. Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368058

Safety Information

signalword Warning
pcodes P411, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KH-0713_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart