KEY410536905 Key Organics / BIONET

3-(4-methoxyphenyl)-2,4-diaza-3-boratricyclo[7.3.1.0^{5,13}]trideca-1(13),5,7,9,11-pentaene

  • MDL number MFCD20527123
  • Cas Number : 1159803-53-8
  • Molecular Weight : 274.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/367304

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2706_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart