KEY410536915 Key Organics / BIONET

2-[4-(dimethylamino)phenyl]-2-hydroxy-1-phenylethan-1-one

  • MDL number MFCD00837364
  • Cas Number : 33458-29-6
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-(dimethylamino)phenyl)-2-hydroxy-1-phenylethanone
Inchi Code 1S/C16H17NO2/c1-17(2)14-10-8-13(9-11-14)16(19)15(18)12-6-4-3-5-7-12/h3-11,16,19H,1-2H3
Inchi Key DGPYFXUBTULRPD-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368078

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4619_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart