KEY410536918 Key Organics / BIONET

methanesulfonic acid amino 4-methoxybenzoate

  • MDL number MFCD11111599
  • Cas Number : 872851-29-1
  • Molecular Weight : 263.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368081

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4644_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart