KEY410536920 Key Organics / BIONET

2,3-dihydro-1,4-benzodioxin-5-amine hydrochloride

  • MDL number MFCD08056380
  • Cas Number : 16081-46-2
  • Molecular Weight : 187.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368083

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4646_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart