KEY410536921 Key Organics / BIONET

9-methyl-3,9-diazabicyclo[4.2.1]nonan-4-one

  • MDL number MFCD08693835
  • Cas Number : 7309-42-4
  • Molecular Weight : 154.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 9-methyl-3,9-diazabicyclo[4.2.1]nonan-4-one
Inchi Code 1S/C8H14N2O/c1-10-6-2-3-7(10)5-9-8(11)4-6/h6-7H,2-5H2,1H3,(H,9,11)
Inchi Key ZPLMTUXXQQUKDH-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368084

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4647_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart