KEY410536925 Key Organics / BIONET

6-bromoimidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD04116918
  • Cas Number : 749849-14-7
  • Molecular Weight : 241.044

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368088

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4651_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart