KEY410536933 Key Organics / BIONET

4,5-dimethyl-1-(triphenylmethyl)-1H-imidazole

  • MDL number MFCD26523312
  • Cas Number : 23593-70-6
  • Molecular Weight : 338.454

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368095

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4659_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart