KEY410536945 Key Organics / BIONET

(but-3-en-1-yloxy)(tert-butyl)diphenylsilane

  • MDL number MFCD26523314
  • Cas Number : 135006-32-5
  • Molecular Weight : 310.512

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368106

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4671_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart