KEY410536955 Key Organics / BIONET

tert-butyl 5-oxo-octahydrocyclopenta[c]pyrrole-2-carboxylate

  • MDL number MFCD11616063
  • Cas Number : 146231-54-1
  • Molecular Weight : 225.288

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368115

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4681_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart