KEY410536976 Key Organics / BIONET

6-bromo-4-methoxy-1H-indazole

  • MDL number MFCD07781361
  • Cas Number : 885519-21-1
  • Molecular Weight : 227.061

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368136

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-6020_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart