KEY410536993 Key Organics / BIONET

5-bromo-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD09701292
  • Cas Number : 552330-86-6
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 263 - 267
Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368153

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-6037_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart