KEY410537008 Key Organics / BIONET

5-iodopyrimidine

  • MDL number MFCD00233961
  • Cas Number : 31462-58-5
  • Molecular Weight : 205.986

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368168

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-6053_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart