KEY411785477 Key Organics / BIONET

potassium trifluoro({4'-methoxy-[1,1'-biphenyl]-4-yl})boranuide

  • MDL number MFCD26936350
  • Molecular Weight : 290.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368193

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2729_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart