KEY411785483 Key Organics / BIONET

N-(4-formyl-2-nitrophenyl)acetamide

  • MDL number MFCD00092015
  • Cas Number : 51818-98-5
  • Molecular Weight : 208.173

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368199

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2735_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart