KEY411785487 Key Organics / BIONET

4-chloro-1H-pyrazol-3-amine

  • MDL number MFCD03419764
  • Cas Number : 54301-34-7
  • Molecular Weight : 117.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119 - 120
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/370732

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FH-0225_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart