KEY411785490 Key Organics / BIONET

6-hydroxy-5-methylpyridine-3-carbaldehyde

  • MDL number MFCD21087785
  • Cas Number : 1289194-02-0
  • Molecular Weight : 137.138

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230 - 232
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/370734

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LH-0725_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart