KEY411785502 Key Organics / BIONET

ethyl 2-(1,3-dithietan-2-ylidene)-3-oxobutanoate

  • MDL number MFCD18800140
  • Cas Number : 120596-96-5
  • Molecular Weight : 218.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368212

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4688_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart