KEY411785506 Key Organics / BIONET

methyl 4-amino-3-fluoro-5-nitro-2-(phenylamino)benzoate

  • MDL number MFCD09909911
  • Cas Number : 606093-58-7
  • Molecular Weight : 305.265

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368216

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4692_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart