KEY414334563 Key Organics / BIONET

potassium (3,4-dihydro-2H-1-benzopyran-6-yl)trifluoroboranuide

  • MDL number MFCD18861595
  • Molecular Weight : 240.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402917

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2747_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart